missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protease Inhibitor Tablets, EDTA-free

Catalog No. PIA32966
Change view
Click to view available options
Quantity:
2 Tablets
20 Tablets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA32966 2 Tablets
PIA32965 20 Tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. PIA32966 Supplier Thermo Scientific™ Supplier No. A32966
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Research Use Only. Not for use in diagnostic procedures.

Thermo Scientific™ Pierce Protease Inhibitor Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments

Pierce Protease Inhibitor Tablets protect proteins from proteolytic degradation through inhibition of the serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A. Stable for storage at 4°C for up to 12 months, each mini or large tablet tablet is sufficient for 10mL or 50mL of solution, respectively, and is suitable for reagent preparation prior to protein extraction from tissue and cultured cells.

Highlights:
  • Improved formulation&mdsashtablets dissolve quickly into a clear solution
  • Economical – more cost-effective than other commercially available formulations for equivalent volumes
  • Convenient&mdsashready-to-use, fully disclosed, broad-spectrum formulations
  • Compatible&mdsashuse directly with Thermo Scientific™ Pierce™ BCA Assays
  • EDTA-free- formulated without EDTA, a metalloproteinase inhibitor (only for EDTA-free formulations)

Specifications

Includes 1 vial of inhibitor tablets
Content And Storage 2°C to 8°C
For Use With (Application) protein purification, western blotting
Sufficient For 1 Tablet is Sufficient for 50 mL
Quantity 2 Tablets
Can I use just part of the Pierce Protease Inhibitor XL Capsules, EDTA-Free and save part of the capsule for later use?

We do not recommend “splitting” the capsule into smaller portions. Researchers using less than 500 mL should choose a different format (liquid or tablet) of protease inhibitors.

What formats of protease inhibitors do you offer?

We offer standalone protease inhibitors (AEBSF, Aprotinin, PMSF, EDTA, etc.), as well as cocktails or combinations of protease inhibitors in liquid, tablet, and larger capsule formats. These cocktails of protease inhibitors can also be purchased in combination with a cocktail of phosphatase inhibitors, in both liquid and tablet form, for even greater convenience.

Do you offer any of the components of the Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets individually?

Yes, the following components of the Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets are offered separately:

- AEBSF (Cat. No. 78431)
- Aprotinin (Cat. No. 78432)
- Bestatin (Cat. No. 8433)
- E-64 (Cat. No. 78434)
- Leupeptin (Cat. No. 78435)
- Pepstatin A (Cat. No. 78436)
- EDTA (Cat. No. 17892).

What are the concentrations of the various components of the protease and phosphatase inhibitor tablets and mini tablets?

The final concentration of each component in the protease and phosphatase inhibitor tablets and mini tablets is proprietary. We release only the identities of the inhibitors.

Can I store the reconstituted solution of protease, phosphatase and (protease and phosphatase) inhibitor tablet or mini tablet and if so, for how long?

The Pierce protease, phosphatase, as well as (protease and phosphatase) inhibitor tablets and mini tablets are stable for at least one year when stored at 4 degrees C, but we only recommend preparing fresh solutions just before needed. If you are interested in liquid protease or phosphatase inhibitors, please use our inhibitor cocktails (https://www.thermofisher.com/search/results?query=protease%20inhibitor%20cocktails&focusarea=Search%20All).

Which protein assay is compatible with the protease, phosphatase, as well as (protease and phosphatase) inhibitor tablets and mini tablets?

The Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets are compatible with common protein assays such as BCA Protein Assay, 660 nm Protein Assay, Coomassie Plus (Bradford) Assay, and Detergent Compatible Bradford Assay.

Which protease and phosphatase inhibitor tablets do you offer?

The Pierce protease, phosphatase, as well as (protease and phosphatase) inhibitors are offered as mini tablets, with or without EDTA. Each mini tablet is sufficient for 10 mL of solution and is suitable for reagent preparation prior to protein extraction for cultured cells and tissues. In addition, the Pierce protease inhibitor tablets are offered with or without EDTA and each tablet is sufficient for 50 mL of solution.

Here are the catalog numbers:
-Pierce Protease Inhibitor Mini Tablets (Cat. No. A32953)
-Pierce Protease inhibitor Mini Tablets, EDTA-free (Cat. No. A32955)
-Pierce Phosphatase Inhibitor Mini Tablets (Cat. No. A32957)
-Pierce Protease and Phosphatase Inhibitor Mini Tablets (Cat. No. A32959)
-Pierce Protease and Phosphatase Inhibitor Mini Tablets, EDTA-free (Cat. No. A32961)
-Pierce Protease Inhibitor Tablets (Cat. No. A32963)
-Pierce Protease Inhibitor Tablets, EDTA-free (Cat. No. A32965)


For Research Use Only. Not for use in diagnostic procedures.