missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protease Inhibitor Mini Tablets

Catalog No. p-7078344
Change view
Click to view available options
Includes:
EDTA
EDTA Free
Catalog No. Includes
PIA32954 EDTA
PIA32956 EDTA Free
2 options
For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PIA32954

Supplier: Thermo Scientific™ A32954

Only null left
For Research Use Only. Not for use in diagnostic procedures.

Thermo Scientific™ Pierce Protease Inhibitor Mini Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments

Pierce Protease Inhibitor Tablets protect proteins from proteolytic degradation through inhibition of the serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A. Stable for storage at 4°C for up to 12 months, each mini or large tablet tablet is sufficient for 10mL or 50mL of solution, respectively, and is suitable for reagent preparation prior to protein extraction from tissue and cultured cells.

Highlights:
  • Improved formulation&mdsashtablets dissolve quickly into a clear solution
  • Economical – more cost-effective than other commercially available formulations for equivalent volumes
  • Convenient&mdsashready-to-use, fully disclosed, broad-spectrum formulations
  • Compatible&mdsashuse directly with Thermo Scientific™ Pierce™ BCA Assays

Specifications

Includes EDTA
Content And Storage 2°C to 8°C
For Use With (Application) protein purification, western blotting
Sufficient For 1 tablet is sufficient for 10 mL
Quantity 2 Tablets
Format Tablet