missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Premium Grade Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate)

Optimize applications where product integrity and risk minimization are paramount our highest quality formulation of this popular amine-to-sulfhydryl crosslinker.

$1691.00 - $10237.80

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable No
Molecular Weight (g/mol) 436.37
PEGylated No
Spacer Arm Length 8.3 Å
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PIPG82085
View Documents
Thermo Scientific™
PG82085
100 mg
Each for $1,691.00
Only null left
 
PIPG82086
View Documents
Thermo Scientific™
PG82086
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Optimize applications where product integrity and risk minimization are paramount with the specially characterized Thermo Scientific™ Pierce™ Premium Grade Sulfo-SMCC, our highest quality formulation of this popular amine-to-sulfhydryl crosslinker.

Thermo Scientific Pierce Premium Grade Sulfo-SMCC is our highest quality formulation of this popular amine-to-sulfhydryl crosslinker, specially characterized for applications where product integrity and risk minimization are paramount.

Compared to the standard grade product, Premium Grade Sulfo-SMCC provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. Sulfo-SMCC is a water-soluble, non-cleavable and membrane-impermeable amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane spacer arm (8.3 angstroms). NHS esters react with primary amines at pH 7-9 to form stable amide bonds. Maleimides react with sulfhydryl groups at pH 6.5-7.5 to form stable thioether bonds. The maleimide groups of Sulfo-SMCC are unusually stable up to pH 7.5 because of the cyclohexane bridge in the spacer arm. Because it contains the hydrophilic sulfonyl moiety, Sulfo-SMCC is soluble up to nearly 10mM in water and many commonly used buffers, thus avoiding the use of organic solvents which may perturb protein structure.

Highlights:
  • Sulfo-SMCC – a water-soluble amine-to-sulfhydryl crosslinker with a stabilizing cyclohexane spacer arm
  • High quality – identity and purity confirmed by several tests, including quantitative NMR
  • Product integrity – enhanced level of testing and characterization compared to standard grade
  • Lot retention – ample supply of past lots retained to ensure future process testing
  • Change management – Change Control Notification (CCN) service
  • Consistent manufacture – batch-specific manufacturing documentation review
Applications:
  • Enzyme labeling of antibodies - both enzyme and antibody specificity can be preserved
  • Create specific bioconjugates via one- or two-step crosslinking reactions
  • Create sulfhydryl-reactive, maleimide-activated carrier proteins for coupling haptens
Specifications

Specifications

Amine-Sulfhydryl
436.37
8.3 Å
No
Pierce
Heterobifunctional
Short (<10 Å)
Water
Yes
No
No
Upon receipt store desiccated at -20°C.
Ambient
Chemical Labeling
Maleimide, Sulfo-NHS Ester
Powder
Standard, Single-use, Premium-grade
Crosslinker
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.