missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Plus Activated Peroxidase Kit

Catalog No. PI31489
Change view
Click to view available options
Quantity:
5 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31489 5 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31489 Supplier Thermo Scientific™ Supplier No. 31489

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Periodate-aldehyde-activated horseradish peroxidase (HRP) enzyme and kit to label primary antibodies to obtain high-activity antibody-HRP conjugates.

Thermo Scientific™ EZ-Link™ Plus Activated Peroxidase Kit contains horseradish peroxidase enzyme that has been aldehyde-activated with periodate to enable high-efficiency labeling and preparation of protein or antibody conjugates.

Other amine-reactive chemistries, like glutaraldehyde, have a tendency to polymerize and produce lower amounts of viable conjugate. Sugar residues present on HRP have been oxidized with periodate to produce aldehydes that react with primary amines. Plus Activated Peroxidase is mixed with the protein to be coupled at a pH compatible with your protein or antibody. After incubating the reaction for one to two hours, the conjugate linkage is reduced and the activated HRP quenched with ethanolamine. The conjugate is purified by desalting or by dialysis. If you are preparing an antibody-HRP conjugate, use the Thermo Scientific Pierce Conjugate Purification Kit (Part No. 44920) to separate unreacted enzyme from the conjugate.

Highlights:

  • Reacts with readily accessible primary amines to form a covalent secondary amine bond
  • Can be stored for at least 12 months at -20°C
  • Consistent conjugation yields reliable conjugates
  • Enzyme activity is 120-200 units/mg

Specifications

Quantity 5 reactions
Content And Storage Sufficient For: 5 enzyme-labeling reactions, each with up to 1 mg of antibody (IgG)
• Plus Activated Peroxidase, 5 x 1 mg
• Sodium cyanoborohydride solution, 1 x 0.5 mL
• Quenching Buffer, 25 mL
• PBS Pack (makes 500 mL), 1 pack
• Carbonate-Bicarbonate Buffer Pack (makes 500 mL), 1 pack

Store at -20°C.
Product Line EZ-Link
Sufficient For 5 Enzyme Labeling Reactions
Labeling Target Antibodies (General), Proteins (General)
Label or Dye HRP (Horseradish Peroxidase)
Labeling Scale 0.5-1 mg

For Research Use Only. Not for use in diagnostic procedures.