missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce Plant Total Protein Extraction Kit

Catalog No. FERA44056
Change view
Click to view available options
Quantity:
50 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERA44056 50 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERA44056 Supplier Thermo Scientific™ Supplier No. LSGA44056

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Thermo Scientific Pierce Plant Total Protein Extraction Kit effectively extracts protein from all kinds of dry and fresh plant tissue without liquid nitrogen or organic solvents.

Features of the Plant Total Protein Extraction Kit:
• Efficient—obtain excellent protein yields in less than 10 minutes using a single buffer and a spin column
• Versatile—native or denaturing lysis buffers for protein extraction from leaves, stems, and seeds
• Compatible—extracts can be quantified using the BCA Protein Assay Kit or the Rapid Gold BCA Protein Assay Kit

The Pierce Plant Total Protein Extraction Kit is composed of optimized buffers and filter cartridges that allow for efficient and rapid protein extraction in less than 10 minutes. The kit is designed to rapidly extract denatured or native proteins from plant tissues (leaves, seeds, soft stem, roots, etc.). Simply grind the sample in the lysis buffer in the filter cartridge and centrifuge. The protein extracts can be used for applications such as SDS-PAGE, western blotting, immunoprecipitation, affinity purification, and activity assays.

Specifications

Quantity 50 preps
Product Type Plant Total Protein Extraction Kit
Content And Storage
  • Denaturing Lysis Buffer, 25 mL
  • Native Lysis Buffer, 25 mL
  • Protein Extraction Filter Cartridges, 50 Ea.
  • Collection Tubes, 50 Ea.
  • Plastic Rod
Assay Entry Western Blot
Shipping Condition Room Temperature
Product Line Pierce

For Research Use Only. Not for use in diagnostic procedures.