missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Phosphatase Inhibitor Mini Tablets

Catalog No. p-7078345
Change view
Click to view available options
Quantity:
2 Tablets
20 Tablets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA32957 20 Tablets
PIA32958 2 Tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. PIA32957 Supplier Thermo Scientific™ Supplier No. A32957
Only null left
For Research Use Only. Not for use in diagnostic procedures.

Thermo Scientific™ Pierce Phosphatase Inhibitor Mini Tablets provide complete cell lysate and biological sample protection from dephosphorylation with a broad spectrum phosphatase inhibitor formulation.

Pierce Phosphatase Inhibitor Mini Tablets protect phosphoproteins from serine/threonine phosphatases and tyrosine phosphatases. Each Phosphatase Inhibitor Tablet contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate for broad-spectrum inhibition of phosphatase activity. Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution. The Pierce Phosphatase Inhibitor Tablets are suitable for use with any cell or tissue extract to preserve protein phosphorylation for downstream applications.

Highlights:
  • Economical&mdsashmore cost effective than other commercially available formulations
  • Improved formulation&mdsashtablets dissolve quickly into a clear solution
  • Complete&mdsashthe tablets inhibit acid, alkaline, and protein phosphatases
  • Compatible&mdsashuse with homebrew or commercially available cell lysis reagents and Thermo Scientific Protein Assays

Specifications

Product Type Protease Inhibitor Cocktail
Content And Storage 1 Vial, Store at 2°C to 8°C
Shipping Condition Wet Ice
Form Solid
Product Line Pierce
Sufficient For 10 mL Solution
Inhibitors Phosphatase Inhibitor
Quantity 20 Tablets
Composition Sodium Fluoride, Sodium Orthovanadate, Sodium Pyrophosphate, Beta-Glycerophosphate

For Research Use Only. Not for use in diagnostic procedures.