Learn More
Thermo Scientific™ Phenol:Chloroform + Tris Buffer
Perform DNA extractions with this phenol:chloroform:isoamyl alcohol mixture (25:24:1), accompanied by a buffer to equilibrate the solution at either pH 6.7 or pH 8.0.
Specifications
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C. |
|---|---|
| For Use With (Application) | DNA Extraction |
| Product Type | Phenol Chloroform and Tris Buffer |
| Form | Liquid |
Description
Thermo Scientific™ Pierce Phenol:Chloroform is phenol supplied premixed with chloroform:isoamyl alcohol (25:24:1) and accompanying bottle of buffer to equilibrate at either pH 6.7 or pH 8.0.
Thermo Scientific Pierce Phenol-Chloroform is phenol supplied premixed with chloroform:isoamyl alcohol (25:24:1) and accompanying bottle of buffer to equilibrate at either pH 6.7 or pH 8.0.
Highlights:
- Offers excellent, nuclease-free performance
- Phenol is supplied premixed with Chloroform:Isoamyl Alcohol (25:24:1)
- Supplied at pH 6.7 for special, lower pH DNA extractions; for most DNA applications, the alkaline buffer (included) should be added to produce a pH of 8.0
Specifications
| Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C. | |
| DNA Extraction | |
| Phenol Chloroform and Tris Buffer | |
| Liquid |
For Research Use Only. Not for use in diagnostic procedures.