missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phenol:Chloroform + Tris Buffer

Catalog No. PI17908
Change view
Click to view available options
Quantity:
100 mL
400 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI17908 100 mL
PI17909 400 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI17908 Supplier Thermo Scientific™ Supplier No. 17908

Perform DNA extractions with this phenol:chloroform:isoamyl alcohol mixture (25:24:1), accompanied by a buffer to equilibrate the solution at either pH 6.7 or pH 8.0.

Thermo Scientific™ Pierce Phenol:Chloroform is phenol supplied premixed with chloroform:isoamyl alcohol (25:24:1) and accompanying bottle of buffer to equilibrate at either pH 6.7 or pH 8.0.

Thermo Scientific Pierce Phenol-Chloroform is phenol supplied premixed with chloroform:isoamyl alcohol (25:24:1) and accompanying bottle of buffer to equilibrate at either pH 6.7 or pH 8.0.

Highlights:

  • Offers excellent, nuclease-free performance
  • Phenol is supplied premixed with Chloroform:Isoamyl Alcohol (25:24:1)
  • Supplied at pH 6.7 for special, lower pH DNA extractions; for most DNA applications, the alkaline buffer (included) should be added to produce a pH of 8.0

Specifications

Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
For Use With (Application) DNA Extraction
Product Type Phenol Chloroform and Tris Buffer
Quantity 100 mL
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.