missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Peroxide Assay Reagent B

Catalog No. PI23282
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI23282 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI23282 Supplier Thermo Scientific™ Supplier No. 23282

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detect and measure hydrogen peroxide levels (H2O2) in biological samples using an iron (Fe) and xylenol orange (XO) reagent for microplates or tubes.

Thermo Scientific Pierce Peroxide Assay Reagent B is a formulation consisting of 125μM xylenol orange in water with sorbitol. This formulation is one of the component of Quantitative Peroxide Assay Kit used for detecting and measuring hydrogen peroxide levels based on the oxidation of ferrous to ferric ion in the presence of xylenol orange, present in the biological and other liquid samples.

Features of Peroxide Assay Reagent B:

  • Convenient—no proprietary reagents involved and remove the variability, difficulty and added expense of formulating the reagents yourself
  • Aqueous-compatible formulations— the formulation includes sorbitol, which provides enhanced sensitivity with non-lipid samples

    Applications of Pierce Quantitative Peroxide Assay Kits:

  • General hydrogen peroxide (H2O2) measurement in protein samples
  • Quantitation of peroxides in detergents
  • Assessment of the effects of peroxides on cellular activity
  • Determination of protein glycation; Measurement of peroxide accumulation in lipids

    In these assay kits, hydroperoxides convert the Fe2+ to Fe3+ at acidic pH. With the aqueous-compatible formulation, peroxide first reacts with sorbitol, converting it to a peroxyl radical, which in turn initiates Fe2+ oxidation to Fe3+. In the lipid compatible formulation, the peroxide converts the Fe2+ to Fe3+ directly. In a sulfuric acid solution, the Fe3+ complexes with the xylenol orange (XO) dye to yield a purple product having an absorbance maximum at 560nm. Peroxide levels in test samples can be determined by calculation from the know extinction coefficient of the XO-Fe complex or by reference to a standard curve prepared with hydrogen peroxide solution.Most proteins do not interfere with these assays, although some metal chelators may require use of a blank (i.e., control) to mitigate these effects.

    Related Products

    Pierce™ Quantitative Peroxide Assay Kit (Aqueous) (Cat. No. 23280)

    Pierce™ Quantitative Peroxide Assay Kit (Lipid) (Cat. No. 23285)

    Pierce™ Peroxide Assay Reagent A (Cat. No. 23281)

    Pierce™ Peroxide Assay Reagent C (Cat. No. 23283)

  • Specifications

    Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
    Description Pierce Peroxide Assay Reagent B
    Assay Peroxide Assay Reagent B
    Detection Method Absorbance
    Product Line Pierce
    Product Type Quantitative Peroxide Assay
    Specificity Not Target-Specific
    For Use With (Application) Solution-based Detection, Absorbance
    For Use With (Equipment) Spectrophotometer, Microplate Reader
    Quantity 50 mL

    For Research Use Only. Not for use in diagnostic procedures.