missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Maleic Anhydride Activated Plates

Catalog No. PI15102
Change view
Click to view available options
Color:
Clear
Format:
Standard
Strip Plate
Quantity:
5 Plates
5 x 5 Plates
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Format Quantity
PI15102 Clear Strip Plate 5 x 5 Plates
PI15100 Clear Strip Plate 5 Plates
PI15110 Clear Standard 5 Plates
PI15112 Clear Standard 5 x 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI15102 Supplier Thermo Scientific™ Supplier No. 15102

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Pierce Maleic Anhydride Activated Plates allow attachment of amine-containing peptides and other molecules to microplate wells for use in binding assays.

Pierce Maleic Anhydride Activated Plates are useful for immobilizing peptides and other ligands that do not coat efficiently by passive adsorption. Reaction of the maleic anhydride groups with primary amines (–NH2) results in formation of amide bonds that are stable at neutral pH and above. Acidic conditions hydrolyze the bond; therefore, coating reactions are best performed at pH 8 to 9, and the reacted plates are best used at pH greater than 7 for ELISA and other methods.

These Thermo Scientific Pierce Maleic Anhydride Activated 96-Well Plates are clear, 96-well plates that allow attachment of amine-containing peptides and other molecules to microplate wells for use in binding assays.

Features of Maleic Anhydride Activated Plates:
• Spontaneously react with primary amines
• A direct, simple approach to covalently attach molecules to the well surface of an EIA plate
• Maleic anhydride maintains its integrity and availability for coupling for months under dry, room temperature conditions

Specifications

Binding Property Standard Capacity, Covalent
Format Strip Plate
Content And Storage Upon receipt store plates at room temperature. Product shipped at ambient temperature.
Detection Method Colorimetric
Product Line Pierce
Color Clear
Product Type Detection Plate, Immunoassay, ELISA
Material Polystyrene
No. of Wells 96
Surface Treatment Maleic Anhydride
No. per Pack 5 Plates
Plate Blocking None
Quantity 5 x 5 Plates
Show More Show Less