missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ LTQ Velos ESI Positive Ion Calibration Solution

Catalog No. PI88323
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88323 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88323 Supplier Thermo Scientific™ Supplier No. LSG88323

missing translation for 'validMainframeMsg'

Calibrate LTQ Velos, LTQ Orbitrap and other mass spectrometers using a mixture of highly purified ionizable molecules designed for positive mode calibration.

Thermo Scientific™ Pierce LTQ Velos ESI Positive Ion Calibration Solution is a mixture of highly purified ionizable molecules designed for positive mode calibration of Thermo Scientific™ LTQ Velos series mass spectrometer instruments.

The Pierce LTQ Velos ESI Positive Ion Calibration Solution is a ready-to-use liquid formulation ideal for quickly performing the routine calibration required to maintain the robust performance of Thermo Scientific Mass Spectrometers. The LTQ Velos ESI Positive Ion Calibration Solution is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.

Highlights:

  • Strong peaks – mixture of caffeine, MRFA, Ultramark 1621 and n-butylamine in a acetonitrile/methanol/acetic solution
  • Ready-to-use – load the mass reference standard into a syringe and inject into the instrument
  • High-purity – mass spectrometry grade reagents in a non-leachable container
  • Stable – store at room temperature for more than one year

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Calibration Solution
Product Line Pierce
Content And Storage Store between 2°C and 8°C for one year (see CofA for specific expiration date of each lot).
Starting Material Peptides
Final Product Type Peptides
Detection Method Mass Spectrometry
Workflow Step Calibration
Quantity 10 mL
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.