missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein A IgG Plus Orientation Kit, 2 mL

Catalog No. PI44893
Change view
Click to view available options
Quantity:
2 Columns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI44893 2 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI44893 Supplier Thermo Scientific™ Supplier No. 44893

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bind and covalently immobilize antibodies to create a reusable affinity purification column with high binding capacity with Protein A or G agarose columns and DSS crosslinker.

Thermo Scientific™ Pierce IgG Plus Orientation Kits bind and crosslink antibody to Protein A or Protein G agarose resin, thereby creating a reusable affinity column for purification of antigens.

These IgG orientation kits use a crosslinker to covalently attach antibody that is bound to Protein A or Protein G agarose resin, thereby creating a reusable affinity column for purification of antigens. The Plus Orientation Kits provide for more efficient and stable antibody immobilization than the traditional orientation method because they use the NHS-ester crosslinker DSS (disuccinimidyl suberate) rather than the imidoester crosslinker DMP (dimethyl pimelimidate). Several milligrams of IgG can be bound and crosslinked to each 2mL column, afterwhich the prepared affinity column can be used multiple times for antigen purification using typical binding and elution conditions.

The immobilization method is called orientation because antibodies are first captured to the affinity resin through their Fc regions, which are the target binding sites of Protein A and Protein G ligands. Thus, the antibodies are bound with their antigen-binding sites oriented outward. DSS crosslinker reacts with primary amines (side chain of lysine residues) of the Protein A/G and affinity-bound antibody, thereby covalently and permanently attaching the proteins together.

Highlights:

  • Contaminant-free antigen purification – enables antigen purification without contamination (co-elution) of antibody

  • Covalent, leak-resistant antibody immobilization – high-capacity binding of IgG to Protein A or Protein G agarose, followed by efficient crosslinking with DSS crosslinker

  • High-quality agarose resin – crosslinked 6% beaded agarose with recombinant Protein A or Protein G

  • Gentle binding and crosslinking conditions – coupling antibody in standard PBS

  • Oriented immobilization – binds and crosslinks IgG with the antigen-binding sites directed outward, able to capture passing antigen in the mobile phase without steric hindrance

  • Complete kits – include all the reagents needed to immobilize the antibody

  • Better than DMP – less antibody leaching than traditional orientation method with dimethyl pimelimidate crosslinker

Includes:

Protein A or G columns, disuccinimidyl suberate (DSS), PBS pack, blocking buffer, binding/wash buffer, elution buffer, and column accessories

Specifications

Content And Storage Sufficient For: Immobilizing 2 to 10 mg of rabbit IgG per column
• Recombinant Protein A Columns, 2 mL, 2 columns
• Disuccinimidyl Suberate (DSS), 2 x 13 mg
• PBS Pack (makes 500 mL), 1 pack
• Blocking Buffer, 6 mL
• IgG Elution Buffer, 15 mL
• Antibody Binding/Wash Buffer, 120 mL
• Column Accessories

Store at 4°C.
Product Line Pierce
Quantity 2 Columns
Type IgG Plus Orientation Kit
Stationary Phase Protein A
Sufficient For 2 to 10 mg of Rabbit IgG per Column
Format Spin Column
Column Type Affinity, Agarose Resin
Capacity (Metric) 2 mL
What are the Pierce Protein A and Protein G IgG Plus Orientation kits for?

The Orientation kits allow for binding of an antibody to the Protein A or G resin. After the antibody is bound, an amine- reactive crosslinker (DSS) is added to covalently attach the antibody to the support. This allows for the target antigen to elute free of antibody contamination. The Protein A and G Orientation Kits are used for larger-scale purification of IgG than is typical in an immunoprecipitation.

For Research Use Only. Not for use in diagnostic procedures.