Learn More
Thermo Scientific™ Pierce™ High Capacity NeutrAvidin™ Agarose
Description
Thermo Scientific™ Pierce NeutrAvidin Resins provide less nonspecific binding and cleaner results than avidin or streptavidin for many kinds of biotin affinity procedures.
Additional Information and Online Ordering
NeutrAvidin Resins are prepared by covalently coupling NeutrAvidin Protein to the beaded agarose or UltraLink Biosupport using efficient and stable chemistries, resulting in NeutrAvidin Agarose and UltraLink Resins that are leach-resistant and stable at pH 2-11. The products are excellent choices for a variety of small- or large-scale affinity purification applications involving biotinylated macromolecules, including separation of biotinylated molecules from samples and immunoprecipitation of antigens using biotin-labeled antibodies.
Highlights:
Low nonspecific binding – unlike avidin, NeutrAvidin Protein has no carbohydrates, eliminating nonspecific binding problems caused by sugars; unlike streptavidin, NeutrAvidin Protein has no Arg-Tyr-Asp sequence, which can be a source of binding to cell surface molecules
Versatile resins – agarose (polysaccharide-based) and UltraLink Biosupport (acrylamide-based) resins provide two different options for particular experimental conditions and are compatible with gravity-flow, spin and FPLC methods
Available in multiple formats – choose from two resin types, regular or high capacity preparations, and small or large-scale package sizes (bulk sizes are also available)
Superior performance – High Capacity Resin has higher binding capacity than best alternative supplier streptavidin products, allowing use of small amounts of resin for experiments
Item-Specific Features:
- Agarose Resins are crosslinked 6% beaded agarose
- UltraLink Resins are beaded acrylamide support (UltraLink Biosupport)
- Plus and High Capacity Resins have a higher loading of NeutrAvidin Protein than the regular forms
Recommended for:
Immunoprecipitation; Purifying proteins that bind to biotinylated ligands; Capturing biotinylated cell-surface proteins; Purifying biotinylated peptides
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. Do not freeze resin. Product is shipped at ambient temperature. |
| Particle Size | 45 to 165 μm |
| Product Line | NeutrAvidin, Pierce |
| Quantity | 10 mL |
| Target | Biotin-Tagged Molecule |
| Type | Agarose |
| Stationary Phase | NeutrAvidin Protein |
| Format | Bottle |
| Column Type | Affinity, Agarose Resin |
| Binding Capacity | 8 mg Biotinylated BSA Binding/mL Resin |
For Research Use Only. Not for use in diagnostic procedures.