missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Guardian™ Peroxidase Conjugate Stabilizer/Diluent

Catalog No. PI37548
Change view
Click to view available options
Quantity:
200 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI37548 200 mL
PI37552 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI37548 Supplier Thermo Scientific™ Supplier No. 37548

Preserve diluted (1-1,000ng/mL) HRP conjugates for more than six months at room temperature without significant loss of activity using Guardian Peroxidase Conjugate Stabilizer/Diluent.

Thermo Scientific™ Guardian Peroxidase Conjugate Stabilizer/Diluent preserves diluted (1-1,000ng/mL) HRP conjugates for more than six months at room temperature without significant loss of activity.

  • Assay-compatible - simply add your favorite blocking buffer to create the ideal diluent for your HRP-based ELISA system

  • Convenient - store ready-to-use dilutions (1:1,000-1:100,000) that maintain enzyme activity in the refrigerator without aliquoting

Specifications

Chemical Name or Material Peroxidase Conjugate Stabilizer/Diluent
Physical Form Liquid
Product Line Guardian
Quantity 200 mL

For Research Use Only. Not for use in diagnostic procedures.