missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Goat Anti-Mouse IgG Antibody Coated Plates, Clear, 96-Well

Catalog No. PI15134
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
PI15134 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI15134 Supplier Thermo Scientific™ Supplier No. 15134

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates.

Thermo Scientific™ Pierce Antibody Coated Plates are ideal for binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates.

These plates are pre-coated with goat anti-mouse (GAM) or goat anti-rabbit (GAR) IgG polyclonal antibodies, allowing assay-specific antibodies to be captured even from impure protein samples. The plates are supplied pre-blocked and ready-to-use. Pierce Secondary Antibody Coated Plates can be used for direct, indirect, competitive or sandwich assays and are available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.

Highlights:

  • Prevents antibody denaturation caused by direct binding
  • Unlike Protein A or G, these bind only to target species
  • Antibody-binding capacity is higher than direct adsorption
  • Pre-blocked to reduce nonspecific binding
  • Less expensive than Protein A or G plates

Specifications

Binding Property Standard Capacity, Affinity
Content And Storage Note: Plates are activated to 100 μL and supplied pre-blocked with SuperBlock™ Blocking Buffer.

Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag and store desiccated at 4°C.
Detection Method Colorimetric
Packaging 5 Plate
Product Line Pierce
Color Clear
Product Type Detection Plate, Immunoassay, ELISA
Material Polystyrene
No. of Wells 96
Surface Treatment Antibody
Plate Blocking SuperBlock™ Buffer
Show More Show Less