missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fluoraldehyde™ o-Phthaldialdehyde Crystals (OPA)

Numéro de catalogue. pi26015
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PI26015 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PI26015 Fournisseur Thermo Scientific™ Code fournisseur 26015
Il en reste null

Measure peptides or amino acids (Ex 340nm, Em 455nm) with phthalaldehyde, a fluorescent, amine-reactive, HPLC pre- or post-column labeling reagent.

Thermo Scientific™ Pierce Fluoraldehyde (OPA) is o-phthalaldehyde, a highly sensitive fluorescent pre- or post-column derivatization reagent for peptide or amino acid detection and quantitation in HPLC.

Fluoraldehyde Crystals are a specially purified, fluorogenic-grade preparation of o-phthalaldehyde (OPA). This amine-reactive compound is primarily used for high-sensitivity, fluorescent detection of primary amines, including amino acids, peptides, proteins and polyamines in pre- or post-column chromatographic effluents.

Highlights:

  • For pre- or post-column amino acid derivatization for fluorescent detection and quantitation

  • Reacts with all primary amine-containing analytes to yield fluorescent derivatives (ex/em = 340nm/455nm)

  • Provides an accurate measure of both composition and absolute protein-peptide content

  • Ideal for work with recombinant proteins and synthetic peptides

  • Can be used for fluorescent protein or peptide assay

  • Can be prepared as a stable aqueous solution in borate (see instructions)

Reagent Properties:

  • Can be prepared as a stable aqueous solution in borate (see instructions)

  • Alternative names: Fluoraldehyde Crystals, OPA

  • Chemical name: ortho-Phthalaldehyde

  • Reactive toward: Primary amines (–NH2)

  • Chemical formula: C8H6O2

  • CAS number: 643-79-8

  • Molecular weight: 134.12

  • Form: Pale yellow, free-flowing crystals

Spécifications

Product Type o-Phthaldialdehyde
Chemical Reactivity Amine
Product Line Fluoraldehyde
Content And Storage Store at room temperature.
Label or Dye OPA (o-phthaldialdehyde)
Detection Method Fluorescence
Quantity 5 g
Form Solid

For Research Use Only. Not for use in diagnostic procedures.