missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ FlexMix™ Calibration Solution for Auto-Ready Mass Spectrometers

Catalog No. PIA51739
Change view
Click to view available options
Quantity:
2 x 4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA51739 2 x 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIA51739 Supplier Thermo Scientific™ Supplier No. A51739

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A room-temperature stable, ready-to-use liquid formulation for the calibration of Thermo Scientific™ Mass Spectrometers with the integrated Auto-Ready ion source.

  • Extended mass range improves mass accuracy
  • Positive and negative mode calibration, a single formulation with extended mass ranges for both ionization calibration modes
  • High-quality, rigorously-tested formulation with lot-specific certificates of analysis
  • Bottle may be directly loaded into the instrument for scheduled automatic calibration
  • Stable at room temperature for 18 months, and within the auto-ready mass spectrometer for up to 4 months

Specifications

Physical Form Liquid
Color Colorless
Quantity 2 x 4 mL
Percent Purity Mixture
In which mass spectrometer instruments can I use the Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers?

Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers is designed to be used in mass spectrometer instruments with autocalibration feature, such as Orbitrap IQ-X Mass Spectrometer using Orbitrap Tribrid Series 3.5 Instrument Control Software (or later).

Does the Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers need to be diluted?

Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers is a ready to use reagent and there is no need for dilution.

Is there a specific mode in which Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers needs to be run?

Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers can be run in Positive or Negative mode.

What is the difference between Pierce FlexMix Calibration Solution and Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers?

Both the Pierce FlexMix Calibration Solution and Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers have the same 16 components and produce the same spectra. However, the Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers has a different storage solution: LC-MS grade acetonitrilie 87% (v/v) and LC-MS grade water 13% (v/v). The FlexMix Calibration for Auto-Ready Mass Spectrometers is designed to be used only in mass spectrometer instruments with autocalibration feature, such as Orbitrap IQ-X Mass Spectrometer using Orbitrap Tribrid Series 3.5 Instrument Control Software (or later).

For Research Use Only. Not for use in diagnostic procedures.