missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL

Catalog No. PIA44296
Change view
Click to view available options
Quantity:
2 x 2 Plates
2 x 25 Columns
25 Columns
Format:
96-well Filter Plate
Spin Column
Volume (Metric):
96-well
0.5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Volume (Metric)
PIA44296 25 Columns Spin Column 0.5 mL
PIA44297 2 x 25 Columns Spin Column 0.5 mL
FERA44305 2 x 2 Plates 96-well Filter Plate 96-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PIA44296 Supplier Thermo Scientific™ Supplier No. A44296

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Enable fast and efficient removal of non-reacted fluorescent dyes, biotin, reducing agents, and crosslinkers from protein samples with Thermo Scientific Pierce Zeba Dye and Biotin Removal columns and filter plates.

Quickly remove non-reacted fluorescent dyes, biotin, reducing agents, and crosslinkers from protein samples with a 7K MWCO in fewer than 15 minutes with Thermo Scientific Pierce Zeba Dye and Biotin Removal columns and filter plates.

Removing free-dye after a labeling reaction is often difficult and time-consuming but is essential for accurate determination of dye-to-protein ratios. Pierce Zeba Dye and Biotin Removal resin is highly specialized to produce exceptional protein recoveries while effectively removing non-conjugated fluorescent dyes, biotinylation reagents, reducing agents, and crosslinkers. When using the appropriate amount of sample and working within buffer conditions, these columns and filter plates can successfully remove almost any fluorescent dye. Even dilute protein samples (25 μg/mL) can be successfully processed to remove dyes, biotin, reducing agents, and crosslinkers and to efficiently recover proteins (>7 kDa). These products are not recommended for desalting or buffer exchange.

Features of Zeba Dye and Biotin Removal columns and filter plates include:
• Versatile—removes unreacted fluorescent dyes, biotinylation reagents, crosslinkers, and reducing agents from protein solutions
• High recovery—low-binding resin maximizes protein recovery
• Easy-to-use—no cumbersome column preparation or equilibration
• Fast—small-molecule removal and protein recovery in less than 15 minutes
• Flexible—available in spin columns and filter spin plates for a range of needs

Sample capacities
• 0.5-mL spin column: 50 to 120 μL sample
• 2-mL spin column: 400 to 700 μL sample
• 5-mL spin column: 1 to 2 mL sample
• 96-well filter plate: 40 to 100 μL sample (per well)

Available formats
• Spin columns: 0.5, 2, 5, and 10 mL
• 96-well filter plates: pre-dispensed; compatible with centrifugation for manual or automated purification; enable fast, consistent well-to-well and plate-to-plate reproducibility for small-scale, high-throughput clean-ups

Applications
• Ideal for post-reaction clean-up of samples
• To effectively remove non-conjugated dye, biotin, reducing agents, and crosslinkers with exceptional protein recovery

Specifications

Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Room Temperature
Format Spin Column
Column Type Proprietary Resin
Product Line Zeba
Quantity 25 Columns
Volume (Metric) 0.5 mL
Sample Volume (Metric) 50 to 120 μL
Are Zeba Dye and Biotin Removal Spin Columns/Filter Plates compatible with mass spectrometry?

Zeba Dye and Biotin Removal Spin Columns/Filter Plates are not directly compatible with mass spectrometry.

Have you tested the Zeba Dye and Biotin Removal Spin Columns/Filter Plates using fixed angle rotor and swinging bucket rotor centrifuges?

The performance of the Zeba Dye and Biotin Removal resin is not affected by the type of centrifuge rotor.

How dilute can my protein sample be for using with Zeba Dye and Biotin Removal Spin Columns/Filter Plates?

Protein samples with as low a concentration as 25 µg/mL can be successfully processed with Zeba Dye and Biotin Removal Spin Columns/Filter Plates to remove dyes, biotin, reducing agents, and crosslinkers and to efficiently recover proteins (>7 kDa).

When should I use Zeba Dye and Biotin Removal Spin Columns/Filter Plates?

Zeba Dye and Biotin Removal Spin Columns/Filter Plates enable fast and efficient removal of non-conjugated fluorescent dyes, biotin, reducing agents, and crosslinkers from protein samples. We have tested removal of the following:
- Alexa Fluor dyes: 488, 555, 647
- Dylight dyes: 488, 550, 650, 650-4x PEG
- Rhodamine dye
- Biotin
- Reducing agents: DTT, TCEP
- Crosslinker: BS3