missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ DST (disuccinimidyl tartrate)
Description
Thermo Scientific™ Pierce DST, also called disuccinimidyl tartrate, is a water-soluble crosslinker that contains amine-reactive NHS ester ends around a 4-atom spacer arm, whose central diol can be cleaved by periodate oxidation.
Highlights:
- Cleavable by Sodium meta-Periodate
- Ideal for applications in which crosslink cleavability is desired without disturbing protein S-S bonds
- DST is soluble in water, DMF or DMSO at a concentration of ∽10mM
- Reactive groups: NHS ester (homobifunctional)
- Reactive toward: amino groups
Spécifications
Spécifications
| Chemical Reactivity | Amine-Amine |
| Cleavable | By Periodate |
| Description | DST |
| Molecular Weight (g/mol) | 344.23 |
| PEGylated | No |
| Spacer Arm Length | 6.4 Å |
| Content And Storage | Upon receipt store desiccated at 4°C. |
| Cell Permeability | Yes |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Afficher plus de résultats |
For Research Use Only. Not for use in diagnostic procedures.