missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Biotin 3' End DNA Labeling Kit

Catalog No. PI89818
Change view
Click to view available options
Quantity:
20 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI89818 20 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI89818 Supplier Thermo Scientific™ Supplier No. 89818

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biotinylate single-stranded DNA primers for non-radioactive EMSA and other nucleic acid detection methods.

Thermo Scientific™ Pierce Biotin 3' End DNA Labeling Kit is for tagging single-stranded DNA primers with biotin for use in non-radioactive electrophoretic mobility shift assays (EMSA) and other nucleic acid detection methods.

TdT exhibits a substrate preference of single-stranded DNA, but it will label duplex DNA with 3' overhangs and blunt duplexes, albeit with a lower efficiency. The Biotin 3' End DNA Labeling Kit has been optimized to incorporate 1-3 biotinylated ribonucleotides (biotin-11-UTP) onto the 3' end of DNA strands. This labeling strategy has the advantage of localizing the biotin to the 3' end of the probe where it will be less likely to interfere with hybridization or sequence-specific binding of proteins. Biotin-labeled DNA probes can be used to facilitate non-isotopic detection in a variety of applications including electrophoretic mobility shift assays (EMSA), Northern or Southern blots, colony hybridizations, or in situ hybridizations.

Highlights:

  • Non-isotopic labeling eliminates the hassle of hazardous radioactive materials or difficult-to-dispose-of waste
  • 1-3 biotinylated ribonucleotides onto the 3' end of DNA strands for less interference with hybridization or sequence-specific binding of proteins
  • Biotin-labeled probes are stable for more than one year
  • 30-minute labeling procedure is fast and efficient

Includes:

1 mL 5X TdT Reaction Buffer, 50μL Terminal Deoxynucleotidyl Transferase, 100 microliter Biotin-11-dUTP, 140μL Unlabeled Control Oligo, 40μL Biotin-Control Oligo

Specifications

Content And Storage Sufficient For: 20 labeling reactions (each with 5 pmol DNA)
• 1 mL 5X TdT Reaction Buffer (-20°C)
• 5 μL Terminal Deoxynucleotidyl Tranferase (-20°C)
• 1 μL Biotin-11-UTP (≤4°C)
• 14 μL Unlabeled Control Oligo (-20°C)
• 4 μL Biotin-Control Oligo (-20°C)
Format Kit
Labeling Target DNA
No. of Reactions 20
Product Line Pierce
Product Type DNA Labeling Kit
Quantity 20 Reactions

For Research Use Only. Not for use in diagnostic procedures.