missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Direct Magnetic IP/Co-IP Kit

Catalog No. PI88828
Change view
Click to view available options
Quantity:
40 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88828 40 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88828 Supplier Thermo Scientific™ Supplier No. 88828

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Conjugate any antibody or protein to magnetic beads and immunoprecipitate (affinity purify) target protein complexes without IP-antibody interference.

Thermo Scientific Pierce Direct Magnetic IP/Co-IP Kit uses NHS-chemistry to covalently immobilize IP antibodies onto magnetic beads for effective immunoprecipitation and co-immunoprecipitation.

Magnetic IP with NHS chemistry is convenient and allows for the immobilization of IP antibody from any species and subtype without the variations in coupling efficiency seen with Protein A and Protein G. The IP antibody is covalently attached so target proteins or co-IP protein complexes can be eluted and analyzed without antibody contamination. The kit contains an optimized protocol and all buffers and reagents necessary for antibody coupling and high yield IP or co-IP using either manual magnetic stands or automated platforms such as the Thermo Scientific KingFisher Instruments.

Features of the Direct Magnetic IP/Co-IP Kit:

  • Antibody-free IP—antibody is irreversibly attached to the magnetic beads, resulting in negligible co-elution of intact antibody or heavy and light chains with the purified antigen
  • Fast—complete antibody conjugation and immunoprecipitation in less than 4 hours
  • Convenient—the IP/co-IP kit contains magnetic beads and all essential buffers for optimal immunoprecipitation
  • Antibody compatible—use with any antibody species, class or isotype
  • Low nonspecific binding—the bead surface is pre-blocked and all nonreacted NHS-ester groups are fully quenched
  • Protocol-compatible—protein coupling to the beads and immunoprecipitation can be performed manually or by automation (e.g., Thermo Scientific KingFisher Instruments)
  • Scalable—use only the amount of antibody needed for a single IP experiment or prepare a larger quantity of antibody magnetic beads for multiple experiments

The protocol for the Pierce Direct IP/Co-IP Kit conjugates IP antibody to Pierce NHS-Activated Magnetic Beads (Catalog #88826) through NHS-ester chemistry. The antibody-bound beads are then incubated with quenching buffer to block remaining active NHS sites. The prepared antibody-bound beads are incubated with cell lysate or tissue extract that contain the protein antigen of interest, allowing the antigen:antibody complex to form. The beads are washed to remove nonbound material and a low-pH elution buffer is used to dissociate bound antigen from the antibody-bound bead. Neutralization buffer is included to prevent precipitation of the isolated antigen and to ensure protein activity in downstream applications. Lane Marker Sample Buffer is included for preparing samples for SDS-PAGE analysis. The protocol is optimized for 2 to 10μg of IP antibody per assay. For optimal co-IP yields, use 10μg of antibody. The beads can be processed manually using a magnetic stand or by automation with an instrument such as the Thermo Scientific KingFisher Flex.

Traditional IP with Protein A/G Magnetic Beads (Catalog #88804), involving passive, noncovalent coupling of antibody with Protein A/G typically provides higher antigen yields than IP protocols that involve covalent attachment of antibody to magnetic beads through crosslinking or NHS-ester chemistry. Covalent attachment inevitably causes loss of some functional antibody binding sites. To overcome this limitation with NHS-chemistry, it may be necessary to use larger amounts of IP antibody in the direct IP method than in the traditional IP method. The advantage of immunoprecipitation with IP antibody covalently bound to the magnetic bead is Western blots and gels devoid of interfering antibody bands.

Related Products

Pierce™ Direct IP Kit (Catalog #26148)

Specifications

Capacity (Metric) 1 mL
Formulation Slurry: 10 mg/mL, 1% solids
Content And Storage Sufficient For: 40 IP reactions using 25 μL of beads
• Pierce NHS-Activated Magnetic Beads, 1 mL
• IP Lysis/Wash Buffer, 2 x 50 mL
• Elution buffer, pH 2.0, 5 mL
• Lane Marker Sample Buffer, Non-reducing, (5X), 5 mL
• Neutralization Buffer, pH 8.5, 0.5 mL
• 0.67M Borate Buffer, 1 mL
• BupH Borate Buffer Pack, 1 pack
• Quenching Buffer, 25 mL

Store at 4°C.
Assay Affinity Purification
For Use With (Equipment) KingFisher™ Flex, Magnetic Stand
Product Line Pierce
Quantity 40 reactions
For Use With (Application) Pull-Down, Immunoprecipitation (IP)
Target Not Target-Specific
Type Immunoprecipitation/Co-Immunoprecipitation Kit
Sufficient For 40 Reactions
Format Kit
Show More Show Less
Can you provide the shelf-life for Direct Magnetic IP/Co-IP Kit?

Direct Magnetic IP/Co-IP Kit is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

I am using the Pierce Direct Magnetic IP/Co-IP Kit. The magnetic beads formed an aggregate during the coupling process; how do I avoid this?

The proteins on the bead surface may adhere to the plastic tube and to each other. To avoid this, after the quenching step, add a mild surfactant at a final concentration of 0.05% (v/v) ( e.g., Tween-20 Detergent, Cat. No 85113) to the washes

Note: Do not use detergent in the coupling step.

For Research Use Only. Not for use in diagnostic procedures.