missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ D-Luciferin, Monopotassium Salt

Get convenience and high performance at a low cost in firefly luciferase reporter assays with our D-Luciferin, formulated at greater than 99% purity as both monosodium and monopotassium salts.

$202.92 - $1256.28

Specifications

Content And Storage Store at or below -20°C.
Detection Method Bioluminescence
For Use With (Application) ATP Assay, Gene Reporter Assay
Product Line Pierce
Substrate Properties Chemical Substrate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Salt Form Price Quantity  
Catalog Number Mfr. No. Quantity Salt Form Price Quantity  
PI88291
View Documents
Thermo Scientific™
88291
100 mg Monosodium Salt
Each for $202.92
Only null left
 
PI88294
missing translation for 'viewDocuments'
Thermo Scientific™
88294
1 g Monopotassium Salt
Each for $1,256.28
Only null left
 
Description

Description

Thermo Scientific™ Pierce D-Luciferin is synthesized and manufactured to ensure high performance at a low cost, and the reporter is also tested by HPLC to ensure proper chirality and activity.

We are the only supplier to perform chiral-high performance liquid chromatography (HPLC) testing to ensure that the product is not only pure but also in the active form. Our D-Luciferin outshines the competition on many levels.
Specifications

Specifications

Store at or below -20°C.
ATP Assay, Gene Reporter Assay
Chemical Substrate
Bioluminescence
Pierce
Luciferase Substrate
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.