missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ D-Luciferin, Monopotassium Salt
Get convenience and high performance at a low cost in firefly luciferase reporter assays with our D-Luciferin, formulated at greater than 99% purity as both monosodium and monopotassium salts.
Supplier: Thermo Scientific™ 88294
Description
Thermo Scientific™ Pierce D-Luciferin is synthesized and manufactured to ensure high performance at a low cost, and the reporter is also tested by HPLC to ensure proper chirality and activity.
We are the only supplier to perform chiral-high performance liquid chromatography (HPLC) testing to ensure that the product is not only pure but also in the active form. Our D-Luciferin outshines the competition on many levels.Specifications
| Store at or below -20°C. | |
| ATP Assay, Gene Reporter Assay | |
| Pierce | |
| 1 g | |
| Chemical Substrate |
| Bioluminescence | |
| D-Luciferin, Monopotassium Salt | |
| >99% | |
| Monopotassium Salt | |
| Luciferase Substrate |
For Research Use Only. Not for use in diagnostic procedures.