missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Copper Coated High Capacity Plates

Catalog No. PI15146
Change view
Click to view available options
Color:
Black
Clear
White
Format:
Standard
Strip Plate
Detection Method:
Chemiluminescence
Colorimetric
Fluorescence
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Format Detection Method
PI15146 Clear Strip Plate Colorimetric
PI15148 Black Standard Fluorescence
PI15143 Clear Standard Colorimetric
PI15147 White Standard Chemiluminescence
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI15146 Supplier Thermo Scientific™ Supplier No. 15146

missing translation for 'validMainframeMsg'

Thermo Scientific Pierce Copper Coated High Binding Capacity (HBC) Plates use an exclusive coating process to increase the amount of histidine-tagged protein that will bind to the plate surface.

Pierce Copper Coated Plates are ideal for analyzing polyhistidine-tagged fusion proteins by ELISA-based methods. Proteins that contain a succession of several histidine residues at the amino or carboxyl terminus have a strong binding affinity for metals such as copper. Although copper is less discriminating in its binding specificity than Nickel, the higher binding capacity of these plates is ideal for high throughput screening applications that need improved sensitivity and greater dynamic range.

Thermo Scientific Pierce Copper Coated High Binding Capacity (HBC) Plates use an exclusive coating process to increase the amount of histidine-tagged protein that will bind to the plate surface.

Features of Copper Coated High Binding Capacity Plates include:
• Quantitate previously undetectable histidine-tagged proteins
• Wider dynamic range with four-fold greater capacity than regular nickel chelate-coated plate
• Copper chelate provides greater binding capacity than nickel
• The detection limit is 0.1 pg of polyhistidine tagged fusion protein per well
• Pre-blocked for the ultimate convenience

Note: Copper coated plates are for use with purified His-tagged proteins. Typical cell lysates contain non-his-tagged proteins that may bind to copper, which can reduce binding capacity and signal-to-noise ratios. For cell lysates, use Thermo Scientific Pierce Nickel Coated Plates.

Specifications

Binding Property High Capacity, Affinity
Format Strip Plate
Content And Storage Store at 4°C in unopened pouches. Once opened, store unused plates in a resealable bag with desiccant at 4°C.
Detection Method Colorimetric
Packaging 5 Plates
Product Line Pierce
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Surface Treatment Divalent Copper, Metal Chelate
Plate Blocking Blocker™ BSA Buffer
Quantity 5 plates
Show More Show Less
Do Pierce Copper Coated High Capacity Plates, White, 96-Well (Cat. No. 15147) and Pierce Copper Coated High Capacity Plates, Black, 96-Well (Cat. No. 15148) have clear bottoms to be read from below?

Pierce Copper Coated High Capacity Plates, White, 96-Well (Cat. No. 15147) and Pierce Copper Coated High Capacity Plates, Black, 96-Well (Cat. No. 15148) can only be read from the top as they have opaque bottoms.