missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CT(PEG)12 Carboxy-PEG-Thiol Compound

CT(PEG)n: carboxylic acid and sulfhydryl compound with a 12-unit polyethylene glycol (PEG) spacer; carboxylate and pegylate quantum dots, gold surfaces, silver.

Supplier:  Thermo Scientific™ 26133

Catalog No. PI26133


Only null left
Explore available promotions

Description

Description

Thermo Scientific™ Pierce CT(PEG)12, or carboxy-PEG12-thiol, is a carboxyl- and sulfhydryl-terminated compound that contains a 12-unit polyethylene glycol (PEG) spacer and is used to modify surfaces such as quantum dots, monolayers and magnetic particles.

CT(PEG)12 is monodentate thiol-terminated and carboxylic acid pegylation reagent. The PEG compound has a defined molecular weight and spacer length and is useful for modifying surfaces such quantum dots, self-assembled monolayers and magnetic particles. Functionalization of solid surfaces with this polyethylene glycol reagent provides carboxylate “handles“ for protein immobilization via EDC crosslinking and significantly reduces nonspecific protein binding and provides.

Highlights:

  • Metal-binding – terminal monodentate thiol reacts spontaneously with gold surfaces through dative binding

  • Carboxylate – terminal carboxylic acid group allows conjugation to amine-containing affinity ligands using carbodiimide (EDC) and Sulfo-NHS crosslinking chemistry

  • Polyethylene glycol – PEG groups are flexible, non-immunogenic, hydrophilic, and significantly reduce nonspecific binding of surfaces for protein methods

  • Spacer arm – 12-unit PEG spacer is nearly 48 angstroms long, minimizing steric hindrance for protein immobilization and binding assays

Specifications:

  • Alternative name: Carboxy-PEG12-Thiol
  • Chemical name: 1-mercapto-3,6,9,12,15,18,21,24,27,30,33,36-dodecaoxanonatriacontan-39-oic acid
  • Chemical formula: C27H54O14S
  • Molecular weight: 634.77
  • Spacer arm: 47.8 angstroms
  • Form: colorless to pale yellow liquid
  • Solubility: dissolve in DMSO or DMF for use
Specifications

Specifications

Amine
Conjugating Groups, Polyethylene Glycol (PEG)
12 ethylene glycol units
100 mg
Thiol Compound
Store at -20°C protected from moisture. Shipped at ambient temperature.
Carboxylic Acid, Thiol
Solid
DMF or DMSO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.