missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ C18 Spin Tips & Columns

Catalog No. PI89873
Change view
Click to view available options
Quantity:
25 Columns
50 Columns
96 Tips
96 x 10 μL Tips
96 x 100 μL Tips
Product Type:
Column
Pipette Tip
Spin Tip
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PI89873 50 Columns Column
PI89870 25 Columns Column
PI84850 96 Tips Spin Tip
PI87782 96 x 10 μL Tips Pipette Tip
PI87784 96 x 100 μL Tips Pipette Tip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PI89873 Supplier Thermo Scientific™ Supplier No. 89873

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific Pierce C18 Spin Columns (10–150 μL) and Spin Tips (10–100 μL) effectively remove interfering contaminants, while also maintaining excellent binding and recovery of peptide samples for use upstream of mass spectrometry and other high sensitivity detection methods.

C18 Spin Tips for Peptide Desalting and Cleanup

Pierce™ C18 spin tips are designed for rapid, reproducible peptide desalting, cleanup, and concentration prior to mass spectrometry analysis. Utilizing reversed-phase C18 chemistry, these tips help enable efficient peptide binding and recovery while removing salts, detergents, and other contaminants that can interfere with downstream applications.

Used in combination with Pierce Spin Tip Adaptors (Catalog No. A40011776) and low protein binding microcentrifuge tubes (Catalog No. 90410), C18 spin tips support a simple centrifugation-based workflow that delivers consistent and reliable results across a wide range of sample inputs.

Features of C18 Spin Tips

  • Reliable – proven reversed-phase C18 chemistry
  • Efficient – effectively removes interfering contaminants to improve spectral quality
  • Streamlined – simple workflow using tip adaptor and centrifugation steps

Applications

  • Peptide desalting prior to LC-MS analysis
  • Cleanup following protein digestion workflows
  • Removal of salts, buffers, and contaminants
  • Peptide concentration and sample preparation

How to Use

C18 spin tips use a bind–wash–elute workflow performed by centrifugation. Tips are placed into a tip adaptor seated in a microcentrifuge tube for each step. Simply activate and equilibrate prior to adding your acidified sample. To enhance peptide binding repeat the sample loading step one time. Lastly, wash 2-3 times and elute into a fresh tube using an acetonitrile solution. Refer to Table 1 in the Figures for recommended buffers, volumes and spin speeds for each tip size.

Specifications

For Use With (Equipment) Microcentrifuge, Mass Spectrometer
Product Type Column
Product Line Pierce
Content And Storage Store at room temperature.
Starting Material Peptides, Protease-digested Protein
Final Product Type Peptides
Detection Method Mass Spectrometry
Format Spin Column
Workflow Step Sample Clean Up
Quantity 50 Columns
Sorbent C18
Show More Show Less
Do I need to do C18 clean-up of my mass spectrometry sample before phosphopeptide enrichment?

Yes. C18 clean-up is necessary to remove small molecules that may interfere with phosphopeptide enrichment. Drying after clean-up is also necessary to concentrate the dilute samples and remove C18 organic eluting solvent so samples can be dissolved in the optimized buffers for phosphopeptide binding. Alternative binding buffers are not recommended as phosphopeptide yield and specificity will be affected. For maximal performance, ensure that the clean, dried peptide samples are completely solubilized in binding buffer with agitation for at least 10 min.