missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Horseradish Peroxidase, Biotinylated

Catalog No. PI29139
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI29139 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI29139 Supplier Thermo Scientific™ Supplier No. 29139

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Purified biotin and biotinylated BSA, HRP, AP or FITC for use as controls or to amplify signal in IHC via the avidin-biotin complex (ABC).

Thermo Scientific™ Pierce Biotinylated Proteins include biotin-labeled horseradish peroxidase (B-HRP), alkaline phosphatase (B-AP) and fluorescein (B-FITC) for use as controls or signal amplification in IHC via avidin-biotin complex (ABC) techniques.

Biotinylated HRP and biotinylated AP are most commonly used in immunohistochemistry (IHC) to amplify the signal of biotinylated primary antibodies using the ABC staining method. Biotinylated Fluorescein (FITC) cannot be used to polymerize and amplify the signal to the same degree as biotinylated enzymes because each fluor molecule is tagged with only one biotin molecule. However, the fluorescent variant of traditional ABC staining is possible with this unique molecule.

Highlights:

  • Biotinylated enzymes – biotin-labeled horseradish peroxidase (B-HRP), alkaline phosphatase (B-AP) and beta-galactosidase (B-bGal) for use in avidin-biotin complex (ABC) staining

  • Biotinylated FITC – amplify the fluorescein signal of biotinylated primary antibodies by using avidin or streptavidin plus biotinylated fluorescein (B-FITC)

  • Biotinylated controls – use biotinylated bovine serum albumin (B-BSA) or any one of these molecules as positive controls in biotin quantitation methods

Specifications

Content And Storage Store between 2°C and 8°C.
Form Solid
Product Line Pierce
Label Type Biotin & Analogs
Quantity 5 mg
Are there any His-tags on Pierce Horseradish Peroxidase, Biotinylated (Cat. No. 29139)?

This biotinylated Horseradish Peroxidase is a native protein with no His-tags.

For Research Use Only. Not for use in diagnostic procedures.