missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Anti-HA Magnetic Beads

Catalog No. PI88837
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88837 5 mL
PI88836 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI88837 Supplier Thermo Scientific™ Supplier No. 88837

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform immunoprecipitation of recombinant HA-tagged proteins expressed in bacterial or mammalian cells or in vitro systems, using manual or robotic magnetic separators.

Thermo Scientific Pierce Anti-HA Magnetic Beads are affinity particles for immunoprecipitation of recombinant HA-tagged proteins expressed in bacterial or mammalian cells or in vitro systems, using manual or robotic magnetic separators.

Features of Anti-HA Magnetic Beads:

  • Specific—highly specific anti-HA monoclonal antibody (clone 2-2.2.14) enables high yield and high purity immunoprecipitation
  • Convenient and fast—product instructions provide an easy-to-follow, optimized protocol for immunoprecipitation in approximately one hour
  • Low non-specific binding—stable, pre-blocked beads and specific antibody minimize off-target binding for HA-tag IP or co-IP experiments
  • Versatile—beads are compatible with manual and automated workflows (e.g., Thermo Scientific KingFisher Instruments)

The blocked magnetic bead surface is coated with anti-HA antibody, a highly specific mouse IgG1 monoclonal antibody that recognizes the HA-epitope tag (YPYDVPDYA) derived from the human influenza hemagglutinin (HA) protein. The Pierce Anti-HA Magnetic Beads can be used manually with a magnetic stand, as well as with automated platforms, such as the Thermo Scientific KingFisher Instruments, for high-throughput workflows.

Specifications:

Product Details:

Pierce Anti-HA Magnetic Beads are convenient for the immunoprecipitation (IP) of recombinant HA-tagged proteins and the co-immunoprecipitation (co-IP) of their interacting proteins. The beads are incubated with a cell lysate containing HA-tagged protein and the fusion protein is captured. The beads are subsequently washed and then the target proteins are eluted using 0.1M glycine, pH 2.0, 50 mM NaOH, or SDS-PAGE sample buffer. If gentler elution conditions are desired, 2 mg/mL Pierce HA peptide can be used. The protocol has been optimized for each of these conditions. Anti-HA antibody can be used to detect HA-tagged protein by Western blot analysis.

Specifications

Capacity (Metric) 5 mL
Concentration Slurry: 10mg/mL, 1% solids
Content And Storage Store at 4°C.
Particle Size 1 μm
Form Liquid
Product Line Pierce
Quantity 5 mL
Protein Form Recombinant
Target HA
Type Magnetic Affinity Bead
Ligand Type Antibody
Show More Show Less
Are the Pierce Anti-HA Magnetic Beads (Cat. No. 88836, 88837) compatible with sodium dodecyl sulfate? If yes, up to which concentration?

Yes, the Pierce Anti-HA Magnetic Beads (Cat. No. 88836, 88837) are compatible with sodium dodecyl sulfate, but the concentration should not exceed 0.5%.

What type of recombinant protein purification resins do you offer?

We offer a variety of resins targeting His-tagged fusion proteins or GST fusion proteins. Please see this selection table to choose the right resin for your experiments. Table available at http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-purification-isolation/protein-purification/recombinant-protein-purification.html

We also offer resins (both agarose and magnetic bead-based) with immobilized anti-c-Myc and anti-HA antibodies for targeting c-Myc and HA-tagged proteins respectively. Please check out the following links for the specific products:

-Anti-c-Myc agarose (https://www.thermofisher.com/order/catalog/product/20169?ICID=search-product)
-Anti-c Myc magnetic beads (https://www.thermofisher.com/order/catalog/product/88843?ICID=search-product)
-Anti-HA agarose (https://www.thermofisher.com/order/genome-database/antibody/HA-Epitope-Tag-Antibody-clone-2-2-2-14-Monoclonal/26181)
-Anti-HA magnetic beads: (https://www.thermofisher.com/order/catalog/product/88837?ICID=search-product)

For Research Use Only. Not for use in diagnostic procedures.