missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ 96-Well Polystyrene Plates, Corner Notch

Catalog No. PI15041
Change view
Click to view available options
Quantity:
100 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI15041 100 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI15041 Supplier Thermo Scientific™ Supplier No. LSG15041

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bind antibodies with high capacity using these specially formulated polystyrene plates for ELISA and other immunoassays.

Thermo Scientific™ Pierce 96-Well Microplates are made of specially formulated polystyrene and treated to produce high-capacity antibody-binding characteristics.

Pierce 96-Well Microplates are standard flat-bottom clear and white assay plates for ELISA and other protein-based methods. Each lot is certified to ensure adherence to strict performance criteria. Clear and opaque white varieties are available in 100-plate and 25-plate cases, respectively. Use the clear plates to perform colorimetric ELISA and other assays for measurement in standard spectrophotometer plate readers. Use the white plates to perform fluorescent or chemilumiescent assays for detection in fluorescence or luminometer readers.

Highlights:

  • Specially formulated optically clear polystyrene and treated to provide protein coating capacity
  • High binding capacity surface improves the overall performance in immunoassay applications
  • Certified for performance, having consistently low coefficients of variation (CV) among wells

Specifications

Binding Property Adsorption, Hydrophilic
Content And Storage Store at room temperature.
Description Pierce 96-Well Polystyrene Plates, Corner Notch
Detection Method Colorimetric
Packaging 100 Plate
Product Line Pierce
Color Clear
Product Type Well Plate
Material Polystyrene
No. of Wells 96
Surface Treatment Polystyrene
Well Shape Flat Bottom (F-well)
Plate Blocking None
Quantity 100 plates
Show More Show Less
What kind of 96-well plate do I need for my fluorescent assay?

Select a microplate that is compatible with your instrument. Refer to the manufacturer's manual for types of microplates that are recommended for use with your instrument.

Various types of plastic may autofluoresce. Select a microplate that is rated for fluorescence-based assays and exhibits little or no autofluorescence in the emission range of the reagent you wish to use.

Clear plates are routinely used for fluorescence-based assays, but opaque-walled microplates reduce bleed-through between wells. If you are using live cells, you may need to consider special substrate treatments for the bottom of the wells, such as poly-L-lysine.

For Research Use Only. Not for use in diagnostic procedures.