missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ In-Line Caps for 1.1 mL Microtube System
Description
Thermo Scientific™ 1.1mL Microtubes and Racked Systems consist of 1.1mL tall microtubes supplied pre-racked in boxes to match a standard 96-well microplate footprint.
The microtubes resembles 1mL pipette tips except that they have round bottoms. Like a box of 1mL pipette tips, the racked microtubes match multi-channel pipettors, providing convenience for a variety of reagent- and sample-dispensing procedures.
Highlights:
- 8 x 12 format simplifies and speeds applications in which a multi-channel pipettor is used
- Format is also compatible with robotic workstations
- Thermo Scientific Microtube System can be centrifuged
Spécifications
Spécifications
| Product Type | Microtube Cap |
| Description | Pierce 1.1mL Microtube In-Line Caps |
| Quantity | 120 strips |
| Sterility | Sterile |
| Content And Storage | Store at room temperature. |
| For Use With (Equipment) | 1.1 mL Microtubes |
For Research Use Only. Not for use in diagnostic procedures.