missing translation for 'onlineSavingsMsg'
Learn More

Picric Acid, Saturated Aqueous Solution, Spectrum™ Chemical

Catalog No. 18612375
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612375 1 L Amber Glass Bottle
18612374 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18612375 Supplier Spectrum Chemical Mfg Cor Supplier No. P2011LT
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88-89-1 , 7732-18-5

Specifications

CAS Max % 1
Quantity 1 L
Packaging Amber Glass Bottle
Grade Chemical Solution

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.