Learn More
Waters Limited PIC Reagent D4
Numéro de catalogue. 50785938
Encompass
Paired -Ion Chromatography (PIC Reagents) provide an easy solution for analyzing samples containing ionic species. These reagents allow ionic compounds to be separated by reverse-phase chromatography, saving time by allowing simultaneous assay of acids, bases, amphoteric and neutral compounds. PIC D4 is a 5 vial/pkg of dibutylammonium phosphate in a workable UV range in mobile phase 215+ nm.
Numéro de catalogue. 50785938 Fournisseur Waters Limited Code fournisseur WAT085466
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Paired -Ion Chromatography (PIC Reagents) provide an easy solution for analyzing samples containing ionic species. These reagents allow ionic compounds to be separated by reverse-phase chromatography, saving time by allowing simultaneous assay of acids, bases, amphoteric and neutral compounds. PIC D4 is a 5 vial/pkg of dibutylammonium phosphate in a workable UV range in mobile phase 215+ nm.