missing translation for 'onlineSavingsMsg'
Learn More

Phytyl Acetate (cis- and trans- mixture) 88.0+%, TCI America™

Catalog No. P16755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16755G 5 g
P167525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16755G Supplier TCI America Supplier No. P16755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10236-16-5
Molecular Formula C22H42O2
Molecular Weight (g/mol) 338.58
MDL Number MFCD00673238
InChI Key JIGCTXHIECXYRJ-ILWBRPEASA-N
Synonym Acetic Acid 3,7,11,15-Tetramethyl-2-hexadecenyl Ester, 3,7,11,15-Tetramethyl-2-hexadecenyl Acetate
PubChem CID 53425386
IUPAC Name (2E,7R,11R)-3,7,11,15-tetramethylhexadec-2-en-1-yl acetate
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC\C(C)=C\COC(C)=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 338.58
Percent Purity ≥88.0% (GC)
Physical Form Liquid
Chemical Name or Material Phytyl Acetate (cis- and trans- mixture)
RTECSNumber MM0675000
TSCA Yes