missing translation for 'onlineSavingsMsg'
Learn More

Phytonadione Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015679
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015679 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015679 Supplier Merck Emd Millipore Supplier No. PHR10781G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phytonadione belongs to the category of oral anticonvulsant drugs. It protects a newborn from acquiring hemorrhagic diseases by maintaining a normal blood clotting mechanism.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 84-80-0
Density 0.984 g/mL (at 25°C)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C31H46O2
MDL Number MFCD00214063
Melting Point -20°C
Synonym Vitamin K1; Phytomenadione; 2-Methyl-3-phytyl-1,4-naphthoquinone; 3-Phytylmenadione; Phylloquinone; Vitamin K1(20)
Type Certified Reference Material
Quantity 1 g
Formula Weight 450.70
Grade Certified Reference Material
Show More Show Less