missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phytin, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 500 g |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Phytin |
Safety and Handling
| EINECSNumber | (8)-0488 |
| RTECSNumber | NM7530000 |
| TSCA | Yes |