missing translation for 'onlineSavingsMsg'
Learn More

Phytin, TCI America™

Catalog No. P041025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P041025G 25 g
P0410500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P041025G Supplier TCI America Supplier No. P041025G
Only null left

Chemical Identifiers

CAS 3615-82-5
Molecular Formula C12H40CaO48P12
Molecular Weight (g/mol) 1364.167
MDL Number MFCD00082315
InChI Key FLIYQIYWLFARGI-UHFFFAOYSA-N
Synonym Calcium Phytate
PubChem CID 131674909
IUPAC Name calcium;molecular hydrogen;(2,3,4,5,6-pentaphosphonooxycyclohexyl) dihydrogen phosphate
SMILES [HH].[HH].C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O.C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O.[Ca]

Specifications

Color White-Yellow
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Phytin
EINECSNumber (8)-0488
RTECSNumber NM7530000
TSCA Yes