Learn More
Kent Scientific PhysioSuite™ Monitor Module Upgrade with CapnoScan™ End-Tidal CO2 Monitor
Description
Includes
RightTemp module
- Highly sensitive infrared spectroscopy CO2 sensor
- RightTemp module included
- For mice and rats
- Module upgrade
Specifications
Specifications
| For Use With (Application) | Monitor expired CO2 in intubated rodents |
| Type | PhysioSuite™ Monitor Module Upgrade |
| For Use With (Equipment) | CapnoScan End-Tidal CO2 Monitor |
| Includes | RightTemp module |