missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Vanillic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002381
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002381 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002381 Supplier Merck Emd Millipore Supplier No. PHL89819100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 121-34-6
Product Line phyproof
Linear Formula HOC6H3(OCH3)CO2 H
MDL Number MFCD00002551
Melting Point 208°C to 210°C
Synonym 4-Hydroxy-3-methoxybenzoic acid
Type Reference Substance
Quantity 100 mg
Formula Weight 168.15
Percent Purity ≥98% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG