missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Tubeimoside I, Reference Substance, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002443
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002443 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002443 Supplier Merck Emd Millipore Supplier No. PHL8055010MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 102040-03-9
Product Line phyproof
Linear Formula C63H98O29
Synonym Lobatoside H; Tubeimoside A
Quantity 10 mg
Formula Weight 1319.43
Percent Purity ≥95.0% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG