missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ trans-Cinnamic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002337
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002337 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002337 Supplier Merck Emd Millipore Supplier No. PHL89608100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

Chemical Name or Material Reference Substance
CAS 140-10-3
Melting Point 133°C
Flash Point 160°C
Linear Formula C9H8O2
MDL Number MFCD00004369
Quantity 100 mg
Synonym (2 E)-3-Phenyl-2-propenoic acid; trans-3-Phenylacrylic acid
Formula Weight 148.16
Product Line phyproof

phyproof is a registered trademark of PhytoLab GmbH & Co. KG