missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ (-)-α-Thujone, ≥90% (GC), MilliporeSigma™ Supelco™

Numéro de catalogue. 111002003
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111002003 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111002003 Fournisseur Merck Emd Millipore Code fournisseur PHL89286100MG
Il en reste null

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Spécifications

CAS 546-80-5
Product Line phyproof
Density 0.914 g/mL (at 20°C)
Linear Formula C10H16 O
MDL Number MFCD00001313
Synonym (1 S,4 R)-1-Isopropyl-4-methylbicyclo[3.1.0 ]hexan-3-one
Type Reference Substance
Quantity 100 mg
Formula Weight 152.23
Percent Purity ≥90% (GC)
Grade Primary Reference Standard
Afficher plus de résultats Afficher moins de résultats

phyproof is a registered trademark of PhytoLab GmbH & Co. KG