missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Synephrine, ≥98% (HPLC), MilliporeSigma™ Supelco™
Spécifications
Spécifications
| CAS | 94-07-5 |
| Product Line | phyproof |
| Linear Formula | C9H13 NO2 |
| MDL Number | MFCD00002370 |
| Synonym | 1-(4-Hydroxyphenyl)-2-methylaminoethanol; 4-Hydroxy-alpha-(methylaminomethyl)benzyl alcohol |
| Type | Reference Substance |
| Quantity | 100 mg |
| Formula Weight | 167.21 |
| Percent Purity | ≥98% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG