missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Steviol, ≥98% (HPLC), MilliporeSigma™ Supelco™
Spécifications
Spécifications
| CAS | 471-80-7 |
| Product Line | phyproof |
| Linear Formula | C20H30O3 |
| MDL Number | MFCD00210558 |
| Synonym | (4 alpha)-13-Hydroxykaur-16-en-18-oic acid; ent-13-Hydroxykauran-16-en-19-oic acid; NSC 226902 |
| Type | Reference Substance |
| Quantity | 20 mg |
| Formula Weight | 318.45 |
| Percent Purity | ≥98% (HPLC) |
phyproof est une marque déposée de PhytoLab GmbH & Co. KG