missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Procyanidin B2, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002193
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002193 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002193 Supplier Merck Emd Millipore Supplier No. PHL8955210MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 29106-49-8
Product Line phyproof
Linear Formula C30H26O12
MDL Number MFCD01861513
Synonym (-)-Epicatechin (4 beta-8)-(-)-epicatechin; 4,8 ′′-Bi-[(+)-epicatechin]; cis,cis′′-4,8 ′′-Bi(3,3′,4′,5,7-pentahydroxyflavane); Proanthocyanidin B2
Type Reference Substance
Quantity 10 mg
Formula Weight 578.52
Percent Purity ≥90% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG