missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ (-)-Olivil, ≥90% (HPLC), MilliporeSigma™ Supelco™
Spécifications
Spécifications
| CAS | 2955-23-9 |
| Product Line | phyproof |
| Linear Formula | C20H24O7 |
| MDL Number | MFCD20040348 |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 376.40 |
| Percent Purity | ≥90% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG