missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Mogroside III-E, ≥90% (HPLC), MilliporeSigma™ Supelco™
Spécifications
Spécifications
| CAS | 88901-37-5 |
| Product Line | phyproof |
| Linear Formula | C48H82O19 |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 963.15 |
| Percent Purity | ≥90% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG