missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Ginsenoside Rb2, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 11021-13-9 |
| Product Line | phyproof |
| Linear Formula | C53H90O22 |
| MDL Number | MFCD00221755 |
| Synonym | (3 beta,12 beta)-20-[(6-O-alpha-L-Arabinopyranosyl-beta-D-glucopyranosyl)oxy]-12-hydroxydammar-24-en-3-yl 2-O-beta-D-glucopyranosyl-beta-D-glucopyranoside; NSC 308878 |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 1079.27 |
| Percent Purity | ≥90% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG