missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Estragole, ≥90% (GC), MilliporeSigma™ Supelco™

Catalog No. 111002333
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002333 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002333 Supplier Merck Emd Millipore Supplier No. PHL82205100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 140-67-0
Product Line phyproof
Density 0.965 g/mL (at 25°C)
Linear Formula H2 C=CHCH2C6H4 OCH3
MDL Number MFCD00008653
Boiling Point 215°C to 216°C
Synonym 4-Allylanisole; p-Allylphenyl methyl ether; p-Methoxyallylbenzene; Chavicol methyl ether; Estragole; Isoanethole; Methyl chavicol
Type Reference Substance
Quantity 100 mg
Formula Weight 148.20
Percent Purity ≥90% (GC)
Grade Primary Reference Standard
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG