missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Erianin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 95041-90-0 |
| Product Line | phyproof |
| Linear Formula | C18H22O5 |
| MDL Number | MFCD06795132 |
| Synonym | 2-Methoxy-5-[2-(3,4,5-trimethoxyphenyl)ethyl]phenol |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 318.36 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG