missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Cyanidin 3-arabinoside chloride, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002404
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002404 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002404 Supplier Merck Emd Millipore Supplier No. PHL8961410MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 111613-04-8
Product Line phyproof
Linear Formula C20H19 ClO10
MDL Number MFCD27952825
Synonym Cyanidin 3-O-alpha-L-arabinoside chloride; Cyanidin 3-alpha-L-arabinopyranoside; Cyanidin 3-alpha-L-arabinoside; Cyanidin 3-arabinoside chloride
Type Reference Substance
Quantity 10 mg
Formula Weight 454.81
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG